Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JARID1B Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501561056
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-1056 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1056 Supplier Bethyl Laboratories, Inc Supplier No. A301813AT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

KDM5B is a histone demethylase that demethylates ′Lys-4′ of histone H3, thereby playing a central role in histone code. It does not demethylate histone H3 ′Lys-9′ or H3 ′Lys-27′. KDM5B demethylates trimethylated, dimethylated and monomethylated H3 ′Lys-4′. It acts as a transcriptional corepressor for FOXG1B and PAX9. It favors the proliferation of breast cancer cells by repressing tumor suppressor genes such as BRCA1 and HOXA5. In contrast, KDM5B may act as a tumor suppressor for melanoma.

Specifications

Antigen JARID1B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KDM5B
Gene Accession No. Q80Y84, Q9UGL1
Gene Alias RBBP2H1A, PLU1, PLU-1, CT31, lysine K-specific demethylase 5B, JARID1B, PUT1
Gene Symbols Kdm5b
Host Species Rabbit
Immunogen Between 1494 and 1544
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10765, 75605
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.