Learn More
Cayman Chemical JasmonIc AcId methyl ester 5g
SDP
Supplier:  Cayman Chemical 90000595 G
Encompass_Preferred
A non-steroidal mineralocorticoid receptor antagonist (IC50 9.4 nM) selective for mineralocorticoid receptors over glucocorticoid androgen and progesterone receptors (IC50s >10000 nM for all) increases the urinary ratio of sodium to potassium in bilateral adrenalectomized rats at 0.3 1 or 3 mg/kg decreases systolic blood pressure in DOCA-salt hypertensive rats at 3 mg/kg reduces systolic blood pressure urinary protein levels blood levels of BNP kidney and left ventricle weights and glomerulosclerosis as well as increases blood potassium levels in Dahl salt-sensitive rats fed a high-salt diet at 2 mg/kg per day
Catalog No. 50-289-6209
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.