Learn More
Selleck Chemical LLC JBSNF-000088-25mg
SDP
Supplier:  Selleck Chemical LLC S677925MG
Encompass_Preferred
JBSNF-000088 (6-Methoxynicotinamide), an analog of nicotinamide (NA), is a potent inhibitor of Nicotinamide N-methyltransferase (NNMT) with IC50 of 1.8 µM, 2.8 µM, and 5.0 µM for human NNMT(hNNMT), monkey NNMT (mkNNMT) and mouse NNMT (mNNMT), respectively.
Catalog No. 50-238-9597
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.