Learn More
AdipoGen JC-1
SDP
Supplier:  AdipoGen CDXT0046M100
Encompass_Preferred
Chemical. CAS 3520-43-2. Formula C25H27Cl4IN4. MW 652.23. Synthetic. A dual-emission potential-sensitive probe that can be used to measure mitochondrial membrane potential. JC-1 is a green-fluorescent monomer at low membrane potential. At higher potentials, JC-1 forms red-fluorescent "J-aggregates," which exhibit broad excitation and very narrow emission spectra. The ratio of red to green fluorescence of JC-1 is dependent only on membrane potential, and not influenced by mitochondrial size, shape, or density.
Catalog No. 50-256-0031
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.