Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ JC-1, Calbiochem™,

Catalog No. 4202005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-020-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-020-05MG Supplier MilliporeSigma™ Supplier No. 4202005MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cationic, fluorescent, carbocyanine dye that can be used as a ratiometric indicator of mitochondrial membrane potential in cells, tissues, and isolated mitochondria.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47729-63-5
Molecular Formula C25H27Cl4IN4
Molecular Weight (g/mol) 652.223
InChI Key FYNNIUVBDKICAX-UHFFFAOYSA-M
Synonym jc-1, jc1 dye, jc-1 jodide, cbic2 3, 5,5',6,6'-tetrachloro-1,1',3,3'-tetraethylbenzimidazolocarbocyanine iodide, 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide, bis 5,6-dichloro-1,3-diethyl-2-benzimidazole trimethinecyanine iodide, 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachlorobenzimidazolocarbocyanine iodide, 5,6-dichloro-2-3-5,6-dichloro-1,3-diethyl-2-benzimidazolinylidene propenyl-1,3-diethylbenzimidazolium iodide, cbic2
PubChem CID 5492929
ChEBI CHEBI:52097
IUPAC Name 5,6-dichloro-2-[(E)-3-(5,6-dichloro-1,3-diethylbenzimidazol-3-ium-2-yl)prop-2-enylidene]-1,3-diethylbenzimidazole;iodide
SMILES CCN1C2=CC(=C(C=C2[N+](=C1C=CC=C3N(C4=CC(=C(C=C4N3CC)Cl)Cl)CC)CC)Cl)Cl.[I-]

Specifications

Color Red to Orange
Quantity 5 mg
Solubility Information Soluble in DSMO
Formula Weight 652.2
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material JC-1
Recommended Storage -20°C (-4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.