Learn More
Mitegen Block, JCSG Core Suite 3, 96 conditions, 1.7mL volumes
SDP
Catalog No. NC0936461
Encompass_Preferred
The Joint Center for Structural Genomics formatted the JCSG Core Suite screens as a result of analyzing over 500,000 high-throughput crystallization experiments, performed at the JCSG. The 384 crystallization formulations were selected based on the highest hit rates in initial screening.
Catalog No. NC0936461 Supplier Mitegen Supplier No. 1009844
The Joint Center for Structural Genomics formatted the JCSG Core Suite screens as a result of analyzing over 500,000 high-throughput crystallization experiments, performed at the JCSG. The 384 crystallization formulations were selected based on the highest hit rates in initial screening.