Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems JESS TRAINING
SDP

Catalog No. NC2868778 Shop All Novus Products
Encompass

  • Compatible with Jess Automated Western Blot System.
  • Fluorescence detection as primary method; also supports chemiluminescence.
  • Automated process includes protein separation, immunodetection, and washes.
  • Analysis time is 3 hours for fully analyzed results.
  • Sensitivity at picogram level for chemiluminescence detection.
  • Multiplexing capability available with fluorescent detection.
  • Separation range from 12 to 230 kDa.
  • Samples are loaded into a microplate.
  • Designed for introductory training on the Jess system.
  • Outputs qualitative Western blot data and quantitative standard curves.
  • Features in-capillary protein normalization.

Catalog No. NC2868778 Supplier R&D Systems Supplier No. CTC0002

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

  • Compatible with Jess Automated Western Blot System.
  • Fluorescence detection as primary method; also supports chemiluminescence.
  • Automated process includes protein separation, immunodetection, and washes.
  • Analysis time is 3 hours for fully analyzed results.
  • Sensitivity at picogram level for chemiluminescence detection.
  • Multiplexing capability available with fluorescent detection.
  • Separation range from 12 to 230 kDa.
  • Samples are loaded into a microplate.
  • Designed for introductory training on the Jess system.
  • Outputs qualitative Western blot data and quantitative standard curves.
  • Features in-capillary protein normalization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.