Learn More
Kurin Jet (Blood Culture Collection Device)
Description
- Lowers blood culture contamination rate
- Helps achieve <1% contamination rate
- Minimal blood volume waste
- Closed System - All-In-One Device
- Fits BD, bioMerieux & ThermoFisher Blood Culture Bottles
Specifications
Specifications
| Content And Storage | PushButton Safety Needle, Kurin Jet, Vacutainer Shield to fit BM BC Bottles |
| Type | Blood Culture Collection |
| Quantity | 200 per Cs. |
| Includes | PB Needle 23G, BM BC Bottle |