Learn More
Tocris™ JF-NP-26

Description
Specifications
Specifications
| Chemical Name or Material | (7-(Diethylamino)-2-oxo-2H-chromen-4-yl)methyl (2-((3-fluorophenyl)ethynyl)-4,6-dimethylpyridin-3-yl)carbamate |
| CAS | 2341841-03-8 |
| Quantity | 5 mg |
| Molecular Formula | C30H28FN3O4 |
| Formula Weight | 513.56 |
| Physical Form | Solid |
| Color | Pale-yellow |
| Product Type | Caged Raseglurant |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.