Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JH 295
GREENER_CHOICESDP

Catalog No. 43222
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-222 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-222 Supplier Tocris Bioscience™ Supplier No. 4322/2

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irreversible Nek2 inhibitor

JH 295 is an irreversible Nek2 inhibitor (IC50=770 nM). Inhibits Nek2 via alkylation of residue Cys22. Displays 5-fold selectivity for Nek2 over Cdk1. Exhibits no significant effect on Aurora B or Plk1 kinases, bipolar spindle assembly or spindle assembly checkpoint. Cell permeable.

Specifications

Chemical Name or Material N-[(3Z)-3-[(2-Ethyl-4-methyl-1H-imidazol-5-yl)methylene]-2,3-dihydro-2-oxo-1H-indol-5-yl]-2-propynamide
CAS 1311143-71-1
Assay Percent Range >98%
Quantity 2 mg
Molecular Formula C18H16N4O2
Formula Weight Observed MW: 320.35
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.