Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JH-X-119-01 hydrochloride | 2591344-30-6 | MFCD31544349 | 99.1% | 488.93 | C25H21ClN6O3 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10301710MM 1ML

Encompass_Preferred

JH-X-119-01 hydrochloride is a selective interleukin-1 receptor-associated kinase 1 (IRAK1) inhibitor supplied as a high-purity research compound. It is provided as a 10 mM solution in DMSO (1 mL) or as the solid form for use in biochemical and cellular assays and preclinical inflammation models.

  • Selective IRAK1 inhibition for signaling and inflammation studies.
  • High purity (about 99.1%) suitable for research applications.
  • Supplied as 10 mM solution in DMSO (1 ML) and as solid for formulation flexibility.
  • Molecular weight 488.93 and formula C25H21ClN6O3 for analytical reference.
  • Storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (1 month), sealed and protected from moisture.

Catalog No. 50-002-46320


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.