Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JHW 007 hydrochloride
GREENER_CHOICE

Catalog No. p-4816580
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-511-0 10 mg
43-515-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-511-0 Supplier Tocris Bioscience™ Supplier No. 4351/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity dopamine uptake inhibitor

Dopamine uptake inhibitor; displays high affinity for the dopamine transporter (DAT) (Ki = 25 nM compared to 1330 and 1730 for NET and SERT respectively). Suppresses the effects of cocaine administration in a dose-dependent manner and decreases cocaine and methamphetamine self-administration in rats.

Chemical Identifiers

CAS 202645-74-7
Molecular Formula C24H30ClF2NO

Specifications

Quantity 10 mg
Formula Weight 421.95
Purity >99%
Molecular Weight (g/mol) 421.957
Chemical Name or Material JHW 007 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.