Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JIB 04
GREENER_CHOICE

Catalog No. 497210R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-721-0R 10 mg
49-725-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-721-0R Supplier Tocris Bioscience™ Supplier No. 4972/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pan Jumonji inhibitor; active in vivo

JIB 04 is a pan Jumonji histone demethylase inhibitor (IC50 values are 230, 340, 435, 445, 855 and 1100 nM for JARID1A, JMJD2E, JMJD2B, JMJD2A, JMJD3 and JMJD2C respectively). Exhibits negligible activity at PHD2 and TET1. Selectively blocks cancer cell growth in vitro and diminishes tumor growth in H358 and A549 mouse xenograft models in vivo. Prolongs survival in a mouse model of breast cancer.

Specifications

Quantity 10 mg
Product Type JIB 04
Purity >99%
Molecular Weight (g/mol) 308.76
Inhibitors JIB 04

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.