Learn More
Chemglass Life Sciences J-Kem Temperature Controller, Model 210, Type ""T"" (-200C to 250C), Controller Only.
Supplier: Chemglass Life Sciences CG320010
J-Kem Temperature Controller, Model 210, Type "T" (-200C to 250C), Controller Only. The Model 210 has 1200 Watts of power which is sufficient for heating mantles up to 5L in size along with most laboratory ovens and hot plates. Controller has the ability to display temperature without heating. The Model 210 controls temperature to within 0.1C of liquids from 1ul to 5L.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 3.25" H x 5.25" W x 7.25" DPower: 10 Amps @ 120VAC, 1200 Watts
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.