Learn More
Chemglass Life Sciences J-KEM Temperature Controller, Model 250, Controller Only, Type K, -50°C to 1200°C
SDP
Supplier:  Chemglass Life Sciences CG320412
Encompass_Preferred
J-KEM Temperature Controller, Model 250, Controller Only, Type K, -50°C to 1200°C. The Model 250 has 3 power outlets for maximum versatility in research. Two of the outlets are used for heating up to 1800 watts (mantles up to 22L).The third outlet (360 watts), used normally for cooling, is programmable to provide power below, above, or at the entered temperature. This third outlet can also be used to turn on an alarm for an over or under temperature condition. Controller has the ability to display temperature without heating. The Model 250 controls temperature to within ± 0.1ºC of liquids from 1ul to 22L. Available w/ a choice of Type "T", “J” or “K” thermocouple.PLEASE NOTE: CONTROLLERS ARE SUPPLIED COMPLETE WITH 1/8” DIAMETER X 12” LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. Dimensions: 3.25" H x 7.5" W x 9" DPower: Heating Outlets #1 & #2: 15 Amps @ 120VAC, 1800 Watts Cooling/Alarm Outlet #3: 3 Amps @ 12VAC, 360 Watts
Catalog No. 50-958-206
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.