Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JLP/JIP-4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982093
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2093 Supplier Bioss Supplier No. BS12270R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.

Specifications

Antigen JLP/JIP-4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPAG9
Gene Accession No. O60271
Gene Alias 3110018C07Rik; 4733401I23Rik; 4831406C20Rik; AW552012; Cancer/testis antigen 89; c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; CT89; HLC4; HLC6; HLC-6; HSS; Human lung cancer oncogene 6 protein; Jip4; JIP-4; JLP; JNK interacting protein; JNK/SAPK-associated protein; JNK/SAPK-associated protein 2; JNK-associated leucine-zipper protein; JNK-interacting leucine zipper; JNK-interacting protein 4; Jsap2; JSAP2a; Kiaa0516; lung cancer oncogene 4; MAPK8IP4; Max-binding protein; Mitogen-activated protein kinase 8-interacting protein 4; PHET; PIG6; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; Spag9; sperm associated antigen 9; sperm associated antigen 9 variant 5; Sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1; SYD1
Gene Symbols SPAG9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human JLP/JIP-4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9043
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.