Learn More
JMJD1B Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Specifications
Specifications
| Antigen | JMJD1B |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | KDM3B |
| Gene Accession No. | Q7LBC6 |
| Gene Alias | JmjC domain-containing histone demethylation protein 2B; Jumonji domain-containing protein 1B; Nuclear protein 5qNCA; JMJD1B; C5orf7; JHDM2B; KIAA1082; |
| Gene Symbols | KDM3B |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.