Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2B / KDM4B Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993247
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3247 10 μL
50-199-3246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3247 Supplier Active Motif Supplier No. 61222
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.

Specifications

Antigen JMJD2B / KDM4B
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene KDM4B
Gene Accession No. O94953
Gene Alias 4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; Jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; Lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B
Gene Symbols KDM4B
Host Species Rabbit
Immunogen This JMJD2B / kDM4B antibody was raised against a peptide within the C-terminal region of human JMJD2B.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23030
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.