Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2B Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560494
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0494 Supplier Bethyl Laboratories, Inc Supplier No. A301477A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Members of the Jumonji domain 2 (JMJD2) family contain a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. Like other members of the JMJD1 and JMJD2 families, JMJD2B is a histone demethylase, but is reported to be a very poor enzyme. Similar to JMJD1A, JMJD2B is a transcriptional target of the hypoxia-inducible factor (HIF), and hypoxic cells express elevated levels of the JMJD2B mRNA and protein. Both JMJD1A and JMJD2B retain their histone demethylase activity in hypoxic conditions, suggesting that this activity of JMJD2B might play a role in the hypoxic gene expression program.

Specifications

Antigen JMJD2B
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene KDM4B
Gene Accession No. O94953
Gene Alias JMJD2B; jmjC domain-containing histone demethylation protein 3B; jumonji domain containing 2B; jumonji domain-containing protein 2B; lysine-specific demethylase 4B
Gene Symbols KDM4B
Host Species Rabbit
Immunogen Between 625 and 675
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23030
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.