Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNJ 1661010, Tocris Bioscience™

Catalog No. 326210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-6210 10 mg
32-625-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-6210 Supplier R&D Systems Supplier No. 3262/10
This item is not returnable. View return policy

Selective, reversible FAAH inhibitor

Selective, reversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50 = 12 nM). Brain penetrant and active in vivo.

Chemical Identifiers

CAS 681136-29-8
Molecular Formula C19H19N5OS
Molecular Weight (g/mol) 365.46
MDL Number MFCD00209157
InChI Key BHBOSTKQCZEAJM-UHFFFAOYSA-N
Synonym n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide, 1-piperazinecarboxamide, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl-1-piperazinecarboxamide, acmc-20emff, d0w5wg, jnj hplc, n1-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide
PubChem CID 2809273
IUPAC Name N-phenyl-4-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine-1-carboxamide
SMILES O=C(NC1=CC=CC=C1)N1CCN(CC1)C1=NC(=NS1)C1=CC=CC=C1

Specifications

Quantity 10 mg
Formula Weight 365.45
Percent Purity >99%
Chemical Name or Material JNJ 1661010
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.