Learn More
Apexbio Technology LLC JNJ-47965567 1428327-31-4 10mg
SDP
Supplier:  Apexbio Technology LLC B821510
Encompass_Preferred
JNJ-47965567 (CAS 1428327-31-4) is a selective antagonist of the P2X7 receptor with high permeability to the central nervous system It demonstrates strong affinity for both human and rat P2X7 receptors effectively inhibiting P2X7 receptor-mediated signaling pathways involved in neuronal and glial cell regulation JNJ-47965567 is frequently utilized in research to investigate the physiological and pathological roles of P2X7 receptors in neuroinflammatory processes neurodegenerative diseases and other central nervous system disorders using various animal models
Catalog No. 50-000-87621
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.