Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK Mouse anti-Human, Mouse, Rat, Clone: 1A12E1, Proteintech
SDP

Catalog No. 501736643 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6643 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6643 Supplier Proteintech Group Inc Supplier No. 662101IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This protein can be phosphorylated and this antibody recognizes the 46 kDa and 55 kDa bands in western blot.

C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predomit forms are JNK1 and JNK2.

Specifications

Antigen JNK
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1A12E1
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAPK10
Gene Accession No. P45983, P45984, P49185, P49186, P49187, P53779, Q61831, Q91Y86, Q9WTU6
Gene Alias c Jun N terminal kinase 1, JNK, JNK 46, JNK1, JNK1A2, JNK21B1/2, MAP kinase 8, MAPK 8, MAPK8, PRKM8, SAPK1
Gene Symbols MAPK10, MAPK8, Mapk9
Host Species Mouse
Immunogen JNK Fusion Protein Ag21426
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116554, 25272, 26414, 26419, 26420, 50658, 5599, 5601, 5602
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.