Learn More
ECM Bioscience JNK Phospho-Regulation Immunocytochemistry Kit, ECM Biosciences
Supplier:  ECM Bioscience JK7630
Encompass_Preferred
The JNK1 phospho-regulation kit may be used to examine the pattern of phosphorylation of JNK1 at the dual site (Thr-183/Tyr-185) relative to the total JNK1 pattern. The kit includes monoclonal antibodies to JNK1 (C-terminal region) and JNK1 (Thr-183/Tyr-185), as well as Goat anti-mouse secondary reagents conjugated to DyLight 488 or DyLight 594 for detection using green (493Ex/518Em) or red (593Ex/618Em) filter sets. Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms, Ck, F
Catalog No. 50-206-8741
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.