Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JNK2 Monoclonal Antibody (3C3C12E5)
GREENER_CHOICE

Catalog No. PIMA541070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541070 Supplier Invitrogen™ Supplier No. MA541070
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

JNK2 Monoclonal Antibody for Western Blot, IP, ELISA

MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3C3C12E5
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MAPK9
Gene Accession No. P45984
Gene Alias AI851083; c-Jun kinase 2; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK/SAPK alpha; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; Mapk9; mitogen activated protein kinase 9; mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54a; p54-alpha; p54aSAPK; PRKM8; PRKM9; protein kinase, mitogen-activated 9; SAPK; SAPK1; SAPK1a; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Symbols MAPK9
Host Species Mouse
Immunogen Recombinant Human JNK2 proteimn.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5601
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.