Learn More
Description
Aims to give a strict outlined synopsis of all those lines of biology important for instruction at schools, colleges and universities and suitable for working with the microscope.
Teacher Package
- Set of selected prepared microscope slides in plastic box made under rigourous scientific control and with the most up to date techniques
- Set of overhead transparencies, large size, full color
- Set of sketch- and worksheets with drawings for all slides
- Textbook with detailed description of all slides, drawings and transparencies
- Special cardboard box for storing and packing
Student Package
- Set of selected prepared microscope slides in plastic box (the same as the teacher slides)
- Textbook with detailed description of all slides
- Special cardboard box for storing and packing
Specifications
Specifications
| Format | Multi-media Package |
| Product Type | Multimedia Teacher Package: Human Diseases |
| Includes | Set of selected prepared microscope slides, set of overhead transparencies, set of sketch and worksheets, textbook with detailed description of all slides, drawings and transparencies, special cardboard box |
| For Use With (Application) | For interactive learning and teaching in school and education |
| Subject | Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
