Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multimedia - Packages For Teachers and Students
SCI_ED

Catalog No. S06730 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Product Type:
Multimedia Student Package: the Wonderful World in a Drop of Water
Multimedia Teacher Package: Human and Animal Histology
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S06730 Multimedia Student Package: the Wonderful World in a Drop of Water
S06710 Multimedia Teacher Package: Human and Animal Histology
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S06730 Supplier Johannes Lieder GmbH & Co. KG Supplier No. SSE41

missing translation for 'validMainframeMsg'

For interactive learning and teaching in school and education.

Aims to give a strict outlined synopsis of all those lines of biology important for instruction at schools, colleges and universities and suitable for working with the microscope.

Teacher Package

  • Set of selected prepared microscope slides in plastic box made under rigourous scientific control and with the most up to date techniques
  • Set of overhead transparencies, large size, full color
  • Set of sketch- and worksheets with drawings for all slides
  • Textbook with detailed description of all slides, drawings and transparencies
  • Special cardboard box for storing and packing

Student Package

  • Set of selected prepared microscope slides in plastic box (the same as the teacher slides)
  • Textbook with detailed description of all slides
  • Special cardboard box for storing and packing

Specifications

Format Set
Product Type Multimedia Student Package: the Wonderful World in a Drop of Water
Includes 6 microscope slides in Plastic box, brochure with explanatory text, cardboard box
For Use With (Application) For interactive learning and teaching in school and education
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.