Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PREMIUM QUALITY Prepared Microscopy Slide Set: Phanerogams Part I
SCI_ED

Catalog No. S9512L Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Quantity:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S9512L 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S9512L Supplier Johannes Lieder GmbH & Co. KG Supplier No. 595210E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Depictured explanatory guide

Microscopic slides of roots and stems of flowering plants in set of ten.

LIEDER high quality microslides are manufactured in rigorous scientific controled laboratory conditions. Slides sets MICR-O-SLIDE are prepared with Top quality histologically fixed materials and thus meet all scientific requirements. A clear, multicolored representation of all tissue structures can be viewed on application of special staining techniques.

  • Best glasses with fine ground edges
  • Size: 26 x 76mm (1 x 3 in.)
  • Furnished with printed labels
  • Packed in solid plastic box

Set Includes:

  • Bean (Phaseolus), dicot root, T.S.
  • Corn (Zea mays), typical monocot root, T.S.
  • Spruce (Picea), root T.S.
  • Hyacinth (Hyacinthus), root T.S.
  • Lime (Tilia), woody root, T.S.
  • Fir (Abies), T.S. of woody branch with resin ducts
  • Sunflower (Helianthus), typical dicot herbaceous stem, T.S.
  • Sedge (Cyperus), stem with scattered bundles, T.S.
  • Apple (Pyrus malus), one year twig with vascular bundles and cambium, T.S.
  • Grapevine (Vitis), older woody stem with cork and medullary or pith rays, T.S.

Specifications

Type Type: Roots and stems of flowering plants
Quantity 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.