Learn More
Eurofins Discoverx (+)-JQ1, 1 mg. (92-1149)
Supplier:  Eurofins Discoverx 921149
Encompass_Preferred
JQ1 is a chemical compound which is under development as a non-hormonal male contraceptive drug. Its chemical structure is a triazolothienodiazepine, a series originally discovered by Mitsubishi Pharma [WO/2009/084693], related to benzodiazepine derivatives such as etizolam, however JQ1 does not produce sedative or anxiolytic effects and is instead a potent and selective inhibitor of the bromodomain testis-specific protein BRDT, which is essential for chromatin remodeling during spermatogenesis. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.