Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JSRP1 Monoclonal Antibody (VF1c)
GREENER_CHOICE

Catalog No. MA3933 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3933 Supplier Invitrogen™ Supplier No. MA3933
  • $620.00
    $481.65 / Each of 1

    Save $138.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-933 detects the junctional sarcoplasmic reticulum (SR) protein from rabbit and guinea pig; moderately in canine and rat; and weakly in mouse. MA3-933 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~90 kDa protein representing junctional SR in rabbit skeletal muscle extracts. Immunofluorescence staining of junctional sarcoplasmic reticulum with MA3-933 results in an increase in intensity in fast (Type II) skeletal muscle over slow (Type I) skeletal muscle, and does not cross-react with cardiac muscle. The MA3-933 antigen is rabbit skeletal muscle triads.

The junction between the transverse tubules (T-tubules) and the sarcoplasmic reticulum (SR) of skeletal muscle is called the triad. At the triad, dihydropyridine receptors (DHPR) of the T-tubule serve as voltage sensors in excitation-contraction coupling, while ryanodine receptors (RyR), the calcium release channels, exist in the membrane of the terminal cisternae of the SR. It is thought that during slow phase depolarization of the T-tubule, a third protein, triadin (95 kDa) transmits electrochemical signals to the SR through direct interaction with both DHPR and RyR. Another skeletal muscle specific 90 kDa protein has also been localized to the this junction. This protein is phosphorylated by an intrinsic protein kinase in triads suggesting that it may play an important regulatory or structural role in the skeletal muscle triad junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JSRP1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone VF1c
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene JSRP1
Gene Accession No. Q3MI48
Gene Alias 2300003C06Rik; 2310032K21Rik; homolog of mouse skeletal muscle sarcoplasmic reticulum protein JP-45; JP45; JP-45; Jsrp1; Junctional sarcoplasmic reticulum protein 1; Junctional SR; junctional-face membrane protein of 45 kDa homolog; skeletal muscle sarcoplasmic reticulum protein JP-45
Gene Symbols JSRP1
Host Species Mouse
Immunogen Rabbit skeletal muscle triads.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106026119, 612184, 690423, 71912
Target Species Mouse, Rat, Canine, Rabbit, Guinea Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.