Learn More
Selleck Chemical LLC JTC-801 S2722-50mg
SDP
Supplier:  Selleck Chemical LLC S272250MG
Encompass_Preferred
JTC-801 is a selective opioid receptor-like1 (ORL1) receptor antagonist with IC50 of 94 nM weakly inhibits receptors and
Catalog No. 50-311-2597
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.