Learn More
Cayman Chemical JTE-013, 383150-41-2, 5 mg
Supplier: Cayman Chemical 100094585
JTE-013 is a potent, selective S1P2 receptor antagonist that binds to the human and rat receptors with IC50 values of 17 and 22 nM, respectively, (IC50 values >10 µM for human S1P1 and S1P3).{14560} It reverses the inhibitory effects of S1P on cell migration of vascular endothelial cells and smooth muscle cells. Similarly, JTE-013 reverses the inhibition of S1P on invasion and migration of B16 melanoma cells.{14560,14559} JTE-013 inhibits S1P-induced contraction of, as well as cAMP accumulation in, coronary artery smooth muscle cells.{14558,14556}
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.