Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Julolidine hydrobromide, 97%, Thermo Scientific™

Catalog No. AAH3629806
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3629806 5 g
AAH3629822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3629806 Supplier Thermo Scientific Chemicals Supplier No. H3629806

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83646-41-7
Molecular Formula C12H16BrN
Molecular Weight (g/mol) 254.17
MDL Number MFCD00198009
InChI Key KHWBRFVBPGPEOJ-UHFFFAOYSA-N
Synonym julolidine hydrobromide, 2,3,6,7-tetrahedron-1h,5h-pyrido 3,2,1-ij quinoline hydrobromide, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine hydrobromide, labotest-bb lt00451605, 2,3,6,7-tetrahydro-1h-5h-benzo i,j quinolizine, hbr, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline hydrobromide
PubChem CID 12255670
IUPAC Name 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene hydrobromide
SMILES Br.C1CN2CCCC3=C2C(C1)=CC=C3

Specifications

Melting Point 239°C to 242°C
Color Pink
Quantity 5 g
UN Number UN2923
Sensitivity Hygroscopic
Formula Weight 254.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Julolidine hydrobromide
Hazard Category H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.