Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Julolidine 97.0+%, TCI America™
SDP

Catalog No. J000725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J000725G 25 g
J00075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. J000725G Supplier TCI America Supplier No. J000725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 479-59-4
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
InChI Key DZFWNZJKBJOGFQ-UHFFFAOYSA-N
Synonym julolidine, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine, 1h,5h-benzo ij quinolizine, 2,3,6,7-tetrahydro, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline, 2,3,6,7-tetrahydro-1h,5h-benzo i,j quinolizine, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline, 2,6,7-tetrahydro-1h,5h-benzo ij quinolizine
PubChem CID 68069
SMILES C1CC2=C3C(=CC=C2)CCCN3C1

Specifications

Melting Point 37°C
Boiling Point 280°C
Quantity 25 g
Formula Weight 173.26
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Julolidine
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.