Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Jump'n Jacks
SDP

Catalog No. 14727017
Encompass
Change view
Click to view available options
Size:
Medium
Dimensions (L x W):
3.5 x 3.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Dimensions (L x W)
14-727-017 Medium 3.5 x 3.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-727-017 Supplier Bio-Serv™ Supplier No. K3541
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Made of flexible natural rubber
  • Unique shape minimizes loss through cage
  • Ribbed texture promotes natural play activity
  • Promotes good dental health
  • Cagewasher safe
  • Red

Specifications

Product Type Canine Toy
Color Red
Material Rubber
For Use With (Application) Cleaning and conditioning of teeth and gums
Size Medium
Dimensions (L x W) 3.5 x 3.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.