Learn More
JUN Mouse anti-Human, Mouse, Rat, Clone: 1G6B11, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody is raised against a region of human JUN. Both phosphorylated (p-c-Jun) and unphosphorylated forms of c-Jun, with sizes of 42-45 kDa and 36-39 kDa, respectively are obtain in some experiments.
Specifications
Specifications
| Antigen | JUN |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1G6B11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | JUN |
| Gene Accession No. | P05412, P05627, P17325 |
| Gene Alias | Activator protein 1, AP 1, AP1, c Jun, JUN, jun oncogene, P39, Proto oncogene c Jun, Transcription factor AP 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.