Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JunB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1835 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1835 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1835 Supplier Invitrogen™ Supplier No. PA1835
  • $578.86
    $449.65 / Each of 1

    Save $129.21

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-835 detects JunB from mouse cells. PA1-835 has been successfully used in ChiP, Western blot and immunoprecipitation procedures. By Western blot, this antibody detects an ~38 kDa protein representing JunB from both uninduced and induced L929 cells. The PA1-835 immunogen is a synthetic peptide corresponding to residues D(45) P Y R G L K G P G A R G P G P E(61) of mouse JunB. Reconstitute in 200 μL PBS to create a stock of 1 mg/mL.

JunB, a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JunB
Applications ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Junb
Gene Accession No. P09450, P17275
Gene Alias activator protein 1; AP-1; Jun B; jun B proto-oncogene; Junb; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; myD21; Oncogene Jun-B; Transcription factor jun-B
Gene Symbols Junb
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues D(45) P Y R G L K G P G A R G P G P E(61) of mouse JunB.
Purification Method IgG fraction
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16477, 3726
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.