Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JunB Recombinant Rabbit Monoclonal Antibody (SD081-08)
GREENER_CHOICE

Catalog No. PIMA541188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541188 Supplier Invitrogen™ Supplier No. MA541188
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

JunB Recombinant Monoclonal Antibody for Western Blot, IHC (P), ICC/IF

JunB, a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JunB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone SD081-08
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Junb
Gene Accession No. P09450, P17275, P24898
Gene Alias activator protein 1; AP-1; Jun B; jun B proto-oncogene; Junb; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; myD21; Oncogene Jun-B; Transcription factor jun-B
Gene Symbols Junb
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal human JunB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16477, 24517, 3726
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.