Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JunD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1834 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1834 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1834 Supplier Invitrogen™ Supplier No. PA1834
  • $590.00
    $449.65 / Each of 1

    Save $140.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-834 detects JunD from mouse cells. PA1-834 has been successfully used in Western blot and ChIP procedures. By Western blot, this antibody detects proteins from ~37-41 kDa representing JunD from both un-induced and induced mouse NIH 3T3 cells. The PA1-834 immunogen is a synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD. This sequence is comletely conserved between mouse, human and rat. Reconstitute with PBS.

JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JunD
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 3.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene JUND
Gene Accession No. P15066, P17535, P52909
Gene Alias activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D
Gene Symbols JUND
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD.
Purification Method Affinity chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16478, 24518, 3727
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.