Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Jupiter Electronic Pipette
SDPSCI_ED

Catalog No. S82936 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
100 to 1000 μL
30 to 300 μL
5 to 50 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S82936 0.5 to 10 μL
S82937 5 to 50 μL
S82938 30 to 300 μL
S82939 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. S82936 Supplier United Scientific Supplies Supplier No. UNEPV0010P
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Jupiter Electronic Pipette has deluxe features at an affordable price.

  • Versatile United Scientific™ Jupiter Electronic Pipettes provide fast, accurate results using three separate modes: pipetting, stepper dispensing, and diluting.
  • Quickly select between modes, volumes, and speed using easy two-knob operation and a digital display with clear status indicators.
  • Choose the parameter to be adjusted using the blue knob, and dial in precise settings using the black function wheel.
  • Jupiter pipettes use a reliable, high-performance motor to deliver accuracy and repeatability.
  • The ergonomic, lightweight design is comfortable to handle, and the electronic operation protects against repetitive motion injury.
  • Three separate aspirating and dispensing speeds and up to ten stepper counts let users decide the frequency and quantity of samples to dispense.
  • The lithium-ion battery in Jupiter is easily recharged using either the provided AC adapter or USB cable.
  • Also includes a stand that can be mounted on a flat surface or wall.
  • Electronic operation saves time and reduces manual errors.
  • Comfortable, lightweight design.
  • Three operating modes with quick, accurate adjustments using two knobs.
  • Calibrated and tested in compliance with ISO8655-6 / DIN12650.

Specifications

Compatible Tips 10 μL
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color White
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material Plastic
Model UNEPV0010-P
Number of Channels 1
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 0.5 to 10 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.