Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Jurkat Membrane Extract Lysate, Liquid, WB, 100 μg

Catalog No. 502670821
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-0821 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-0821 Supplier Abcam Supplier No. AB30130100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use Jurkat cell membrane lysate prepared for Western blot analysis.

  • Applications: WB
  • Form: Liquid
  • Storage buffer constituents: HEPES, Magnesium chloride, Potassium chloride, Sucrose, 10% Glycerol, 2% Sodium lauryl sulfate, 1.2% 2-Mercaptoethanol, 1% 4-Nonylphenol, branched, ethoxylated, and 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Detection Location In-Blot Detection
Form Liquid
Target Molecule Human
Buffer HEPES, magnesium chloride, potassium chloride, sucrose, 10% glycerol, 2% sodium lauryl sulfate, 1.2% 2-mercaptoethanol, 1% 4-nonylphenol, branched, ethoxylated, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione
Formulation HEPES, Magnesium chloride, Potassium chloride, Sucrose, 10% Glycerol, 2% Sodium lauryl sulfate, 1.2% 2-Mercaptoethanol, 1% 4-Nonylphenol, branched, ethoxylated, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione
Sample Type Cell Lysate
Quantity 100 μg
For Use With (Application) Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.