Learn More
Stoelting™ Just for Mouse™ Stereotaxic Instrument
SDP
Catalog No. 10000205 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Basic
Digital
Digital, Dual
Dual
Motorized
Ultra Precise, Digital, multiple
Ultra-Precise
Ultra-Precise, Digital, single
Ultra-Precise, Dual
Ultra-Precise, Dual, Digital
Animal:
Mouse
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Animal
10-000-205 Dual Mouse
10-000-203 Basic Mouse
10-000-415 Digital, Dual Mouse
10-000-411 Digital Mouse
10-000-412 Motorized Mouse
10-000-414 Ultra-Precise, Digital, single Mouse
10-000-417 Ultra-Precise, Dual, Digital Mouse
10-000-413 Ultra-Precise Mouse
10-000-416 Ultra-Precise, Dual Mouse
10-000-410 Ultra Precise, Digital, multiple Mouse
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 10-000-205 Supplier Stoelting™ Supplier No. 51733
Add to Cart
Add to Cart
Request a Quote
The Just for Mouse™ Stereotaxic Instrument was specifically designed for use with knock-out and transgenic mice.
  • 25 cm X 25 cm footprint
  • Warmer ready base compatible with Stoelting Rodent Warmer System
  • Gas Anesthesia compatible
  • Ear and tooth bar post height adjustment to accommodate animals between 10-75 g
  • Delrin™ ear bars, with 3 types of head holders
  • Triple lead screw in manipulator arm for fast positioning
  • Dual arm capable (2nd Arm is optional)
  • Digital LED display (Digital Only)
  • Software and motor controlled (Motorized Only)
  • Integrated Stereotaxic Atlas (Motorized Only)
  • Accurate to 100 microns (Non-Digital), 10 microns (Digital Only) or 1 micron (Motorized Only)
Specifications
For Use With (Application) Neuroscience
Animal Mouse
Model Dual