Description
- 25 cm X 25 cm footprint
- Warmer ready base compatible with Stoelting Rodent Warmer System
- Gas Anesthesia compatible
- Ear and tooth bar post height adjustment to accommodate animals between 10-75 g
- Delrin™ ear bars, with 3 types of head holders
- Triple lead screw in manipulator arm for fast positioning
- Dual arm capable (2nd Arm is optional)
- Digital LED display (Digital Only)
- Software and motor controlled (Motorized Only)
- Integrated Stereotaxic Atlas (Motorized Only)
- Accurate to 100 microns (Non-Digital), 10 microns (Digital Only) or 1 micron (Motorized Only)
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Animal | Mouse |
| Model | Ultra-Precise, Digital, single |