Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Base Cover for 90-Minute EN Cabinets

Catalog No. 19414060 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
30 gal. EN cabinet
45 gal. EN cabinet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-414-060 30 gal. EN cabinet
19-414-061 45 gal. EN cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-060 Supplier Justrite™ Supplier No. 22636
Add to Cart
Add to Cart

Keeps debris out from underneath cabinets by adding an optional, removeable base cover

  • Made from textured, durable powder-coated steel
  • Conceals integrated transport base on cabinet
  • Prevents dust and debris from collecting underneath cabinet
  • Creates seamless flush-to-floor attractive presentation
  • Yellow

Specifications

Material Steel
For Use With (Equipment) 30 gal. EN cabinet
Surface Coating Powder-coated
Description Prevents collection of dust, debris; Textured, durable powder-coated steel
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.