Learn More
Justrite™ HPLC Manifold Kit for Quick-Disconnect Disposal Cans: Plug Caps
Catalog No. 19160371 Shop All Justrite Safety Group Products
Change view
Click to view available options
Plug caps
Catalog No.
19-160-371 Plug caps
1 options
Catalog No. 19-160-371 Supplier Justrite™ Supplier No. 28176
Economical way to safely collect waste from multiple HPLC machines
Use with FM-approved Justrite™ Disposal Cans with stainless-steel quick-disconnect fittings to feed up to four waste lines from HPLC processes into a single intake fitting.
  • Four plug caps to close unused ports and reduce vapor release
Certifications
FM approved.
Compliance
Safety can (sold separately) with installed manifold meets OSHA and NFPA Code 30 requirements.
Specifications
Material Polypropylene
Product Type Plug Cap
Certifications/Compliance FM, OSHA, NFPA Code 30
Quantity 1 Ea.
For Use With (Application) Safely collect waste from multiple HPLC machines
For Use With (Equipment) Justrite™ disposal cans with stainless steel quick-disconnect fittings