Learn More
Description
- Ideal for use with flammables
- Leakproof, self-closing dispenser valves minimize spills, controls excess
- High-density polyethylene body
- Permits easy, one-hand dispensing of hazardous liquids directly onto work surface
Specifications
Specifications
| Product Type | Dispensing Can |
| Capacity (English) | 4 oz. |
| Capacity (Metric) | 0.19 L |
| Color | White |
| For Use With (Application) | For use with Flammables |
| Material | Polyethylene |
| Includes | Leakproof Brass Dispenser Valves Self-close |
| Diameter (English) Outer | 2.25 in. |
| Diameter (Metric) Outer | 5.72 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.