Description
- Made of vinyl fabric
- Color coded for visibility and easy identification
- Folds tightly and stows easily
- Embedded foam ring top raises containment wall as fluid level increases
Compliance
EPA for containment and spill prevention (SPCC).
Specifications
Specifications
| Capacity (English) | 100 gal. |
| Capacity (Metric) | 379 L |
| Height (English) | 11 in. |
| Height (Metric) | 27.9 cm |