Learn More
Description
- Leakproof, tough vinyl coated fabric provides broad UV and chemical resistance that withstands repeated usage
- Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports
- Lightweight berm foldable to small size for storage in trucks, emergency vehicles and spill kits
- Add optional grates to help reduce contact with contaminated liquids by personnel or equipment
Applications:
- Saddle tank ruptures
- Personnel and small equipment decontamination
- Fuel tank overspill protection
- Hydraulic leaks and breaks
- Transformer storage
- Leaky containers, machinery, and piping
- Overfills from aircraft or locomotive refueling
- Truck, auto, and forklift battery storage
- Spill kit addition
Compliance
Comply with EPA for containment and spill prevention (SPCC).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (English) | 119 gal. |
| Capacity (Metric) | 450 L |
| Material | PVC-coated Fabric |
| For Use With (Application) | Emergency spill response |
| Dimensions (L x W x H) | 4 x 6 x 8 ft. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
