Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ QuickBerm™ Rigid Lock Spill Containment System

Catalog No. 19111073 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
79 gal.
119 gal.
159 gal.
175 gal.
235 gal.
318 gal.
355 gal.
398 gal.
475 gal.
745 gal.
2510 gal.
Dimensions (L x W x H):
4 x 4 x 8 ft.
4 x 6 x 8 ft.
4 x 8 x 8 ft.
8 x 8 x 8 ft.
4 x 6 x 12 ft.
4 x 8 x 12 ft.
6 x 8 x 12 ft.
8 x 10 x 8 ft.
10 x 10 x 12 ft.
12 x 28 x 12 ft.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W x H)
19-111-073 318 gal. 8 x 8 x 8 ft.
19-111-048 175 gal. 4 x 6 x 12 ft.
19-111-049 235 gal. 4 x 8 x 12 ft.
19-111-050 355 gal. 6 x 8 x 12 ft.
19-111-051 475 gal. 6 x 8 x 12 ft.
19-111-052 745 gal. 10 x 10 x 12 ft.
19-111-072 159 gal. 4 x 8 x 8 ft.
19-111-071 119 gal. 4 x 6 x 8 ft.
19-111-070 79 gal. 4 x 4 x 8 ft.
19-111-074 398 gal. 8 x 10 x 8 ft.
19-111-097 2510 gal. 12 x 28 x 12 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19-111-073 Supplier Justrite™ Supplier No. 28566
Add to Cart
Add to Cart

Portable, versatile containment berm deploys quickly for emergency spill response

  • Leakproof, tough vinyl coated fabric provides broad UV and chemical resistance that withstands repeated usage
  • Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports
  • Lightweight berm foldable to small size for storage in trucks, emergency vehicles and spill kits
  • Add optional grates to help reduce contact with contaminated liquids by personnel or equipment

Applications:

  • Saddle tank ruptures
  • Personnel and small equipment decontamination
  • Fuel tank overspill protection
  • Hydraulic leaks and breaks
  • Transformer storage
  • Leaky containers, machinery, and piping
  • Overfills from aircraft or locomotive refueling
  • Truck, auto, and forklift battery storage
  • Spill kit addition

Compliance

Comply with EPA for containment and spill prevention (SPCC).

Specifications

Capacity (English) 318 gal.
Capacity (Metric) 1203 L
Material PVC-coated Fabric
For Use With (Application) Emergency spill response
Dimensions (L x W x H) 8 x 8 x 8 ft.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.