Learn More
Justrite™ QuickBerm™ Spill Containment Systems
GSA_VA
Catalog No. 19111060 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
175 gal.
235 gal.
355 gal.
475 gal.
745 gal.
1075 gal.
1435 gal.
1795 gal.
1910 gal.
2390 gal.
2990 gal.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-111-060 235 gal.
19-111-059 175 gal.
19-111-061 355 gal.
19-111-064 1075 gal.
19-111-062 475 gal.
19-111-063 745 gal.
19-111-065 1435 gal.
19-111-066 1795 gal.
19-111-067 1910 gal.
19-111-068 2390 gal.
19-111-069 2990 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19-111-060 Supplier Justrite™ Supplier No. 28536
Add to Cart
Add to Cart
Portable containment for drums, buckets, and leaky equipment
QuickBerm secondary containment helps to comply with EPA regulations and provides dependable spill control for leak-prone containers and equipment.
  • Single-piece construction, quick and easy setup, no assembly required
  • Quick response under emergency spill conditions
  • Durable modified PVC coated fabric withstands long-term UV exposure, good for outdoor use
  • Inside wall supports allow berm placement virtually flush up against walls or equipment
  • Reinforced CriticalCorner™ design features wraparound diagonal welds for superior strength and fluid-tight reliability
  • Withstands broad range of chemicals including petroleums and most acids, and is durable enough for extended wear
  • Highly visible yellow inside supports alert against potential catch hazards inside while keeping exterior free from trip hazards found in other berms with exterior support systems
Compliance
Comply with EPA for containment and spill prevention (SPCC)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Capacity (English) 235 gal.
Capacity (Metric) 890 L
Type Spill Control System