Description
QuickBerm secondary containment helps to comply with EPA regulations and provides dependable spill control for leak-prone containers and equipment.
- Single-piece construction, quick and easy setup, no assembly required
- Quick response under emergency spill conditions
- Durable modified PVC coated fabric withstands long-term UV exposure, good for outdoor use
- Inside wall supports allow berm placement virtually flush up against walls or equipment
- Reinforced CriticalCorner™ design features wraparound diagonal welds for superior strength and fluid-tight reliability
- Withstands broad range of chemicals including petroleums and most acids, and is durable enough for extended wear
- Highly visible yellow inside supports alert against potential catch hazards inside while keeping exterior free from trip hazards found in other berms with exterior support systems
Compliance
Comply with EPA for containment and spill prevention (SPCC)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (English) | 2390 gal. |
| Capacity (Metric) | 9049 L |
| Type | Spill Control System |